Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdcd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016688.2 | NP_057897.2 | 3, 4 | 1325 | ||
GenBank mRNA | AF083929.1 | - | 3, 4 | 1319 | ||
AK004096.1 | - | 1, 2 | 118 | |||
AK136141.1 | - | 1, 2 | 104 | |||
AK155050.1 | - | 3, 4 | 1290 | |||
BC022772.1 | - | 2, 3 | 362 | |||
DV656988.1 | - | 2, 3 | 338 |