Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdca8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026560.4 | NP_080836.3 | 9 | 999 | ||
GenBank mRNA | AK003755.1 | - | 2 | 145 | ||
AK076065.1 | - | 9 | 956 | |||
AK076422.1 | - | 9 | 989 | |||
AK077717.1 | - | 9 | 952 | |||
AK077920.1 | - | 9 | 922 | |||
AK078404.1 | - | 9 | 952 | |||
AK082800.1 | - | 9 | 964 | |||
AY550907.1 | - | 9 | 989 | |||
BC068181.1 | - | 8 | 913 | 30634239 |