Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifitm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025378.2 | NP_079654.1 | 2 | 483 | ||
GenBank mRNA | AK003407.1 | - | 2 | 484 | ||
AK007919.1 | - | 2 | 472 | |||
AK151890.1 | - | 2 | 449 | |||
AY082484.1 | - | 2 | 452 | |||
AY594690.1 | - | 2 | 499 | |||
BC010291.1 | - | 2 | 435 | 3152819 |