Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp6v1c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025494.3 | NP_079770.2 | 4 | 470 | ||
GenBank mRNA | AB088407.1 | - | 4 | 417 | ||
AK006323.1 | - | 4 | 421 | |||
AK075779.1 | - | 4 | 442 | |||
AK129007.1 | - | 4 | 430 | |||
AK151237.1 | - | 4 | 431 | |||
AK168914.1 | - | 4 | 421 | |||
BC010217.1 | - | 4 | 418 | 3498388 | ||
U13839.1 | - | 4 | 413 |