Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Coprs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025556.3 | NP_079832.1 | 3 | 190 | ||
GenBank mRNA | AK006497.1 | - | 3 | 224 | ||
AK008882.1 | - | 3 | 191 | |||
AK010573.1 | - | 3 | 192 | |||
BC029192.1 | - | 3 | 187 | 2647299 | ||
BC051120.1 | - | 1 | 1351 |