Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2h5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181392.3 | NP_852057.2 | 3 | 305 | ||
GenBank mRNA | AK003547.1 | - | 3 | 294 | ||
AK003579.1 | - | 3 | 289 | |||
AK011281.1 | - | 3 | 290 | |||
AK012257.1 | - | 3 | 367 | |||
AK013225.1 | - | 3 | 269 | |||
AK019011.1 | - | 3 | 260 | |||
BC029238.1 | - | 3 | 281 | 4489531 |