Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ppcdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176831.4 | NP_789801.1 | 4 | 336 | ||
GenBank mRNA | AK036042.1 | - | 4 | 319 | ||
AK038628.1 | - | 4 | 314 | |||
AK046751.1 | - | 1 | 170 | |||
AK078825.1 | - | 4 | 319 | |||
BC004779.1 | - | 4 | 329 | |||
BC052928.1 | - | 3 | 285 | 5255304 | ||
BC089351.1 | - | 3 | 300 | 30603115 |