Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chmp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024190.2 | NP_077152.1 | 1 | 691 | ||
GenBank mRNA | AK012996.1 | - | 1 | 642 | ||
AK151071.1 | - | 1 | 660 | |||
AK167614.1 | - | 1 | 629 | |||
BC002229.1 | - | 1 | 627 | 3491354 | ||
BC034442.1 | - | 1 | 615 | |||
CT010307.1 | - | 1 | 549 |