Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcsh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026572.3 | NP_080848.1 | 4 | 414 | ||
GenBank mRNA | AK003189.1 | - | 499 | |||
AK003788.2 | - | 4 | 411 | |||
AK009543.1 | - | 2 | 178 | |||
AK019983.1 | - | 4 | 415 | |||
AK050373.1 | - | 4 | 427 | |||
AK178705.1 | - | 1 | 38 | |||
BC014745.1 | - | 4 | 418 | 4239181 |