Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013256.2 | NP_001013274.2 | 13 | 2336 | ||
NM_001143790.1 | NP_001137262.1 | 13 | 2336 | |||
NM_001281464.1 | NP_001268393.1 | 13 | 2207 | |||
NM_001281465.1 | NP_001268394.1 | 13 | 2320 | |||
GenBank mRNA | AK038651.1 | - | 13 | 2123 | ||
AK044055.1 | - | 13 | 2115 | |||
AK165906.1 | - | 13 | 2136 | |||
BC014687.1 | - | 4 | 507 | |||
BC060629.1 | - | 9 | 1386 | 6847619 | ||
BC066078.1 | - | 13 | 2151 | |||
BC072625.1 | - | 13 | 2164 | |||
BC147495.1 | - | 13 | 2134 | 9056325 | ||
BC147497.1 | - | 13 | 2134 | 9056354 |