Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tmigd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027025.4 | NP_081301.2 | 3 | 387 | ||
GenBank mRNA | AK005581.1 | - | 3 | 339 | ||
BC038878.1 | - | 2 | 267 |