Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enkur | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027728.2 | NP_082004.1 | 3 | 587 | ||
GenBank mRNA | AK017056.1 | - | 3 | 588 | ||
AY454124.1 | - | 3 | 585 | |||
BC099519.1 | - | 3 | 486 | 30848009 | ||
BC132425.1 | - | 3 | 448 | 40130702 | ||
BC132451.1 | - | 3 | 448 | 40130728 |