Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mro | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017067.1 | - | 1, 2 | 25 | ||
AY243876.1 | - | 4, 5 | 432 | |||
BY088818.1 | - | 3, 4 | 313 | |||
CK335185.1 | - | 3, 4 | 298 |