Gene Symbol: Kremen2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028416.2 | NP_082692.1 | 4 | 498 | ||
GenBank mRNA | AJ457192.1 | - | 4 | 510 | ||
BC120532.1 | - | 4 | 441 | 40129455 | ||
BC125438.1 | - | 4 | 441 | 40129953 |