Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: D3Ertd751e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099785.1 | NP_001093255.1 | 6 | 622 | ||
NM_001291048.1 | NP_001277977.1 | 4 | 709 | |||
NM_027271.1 | NP_081547.1 | 5 | 487 | |||
NM_028667.3 | NP_082943.2 | 7 | 739 | |||
GenBank mRNA | AK012700.1 | - | 4 | 668 | ||
AK015146.1 | - | 6 | 693 | |||
AK039725.1 | - | 6 | 586 | |||
AK044548.1 | - | 4 | 325 | |||
AK045813.1 | - | 6 | 586 | |||
AK085404.1 | - | 6 | 620 | |||
AK138405.1 | - | 8 | 844 | |||
AK144064.1 | - | 6 | 586 | |||
BC062244.1 | - | 5 | 447 | 1345984 |