Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thyn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144543.2 | NP_653126.1 | 3 | 310 | ||
GenBank mRNA | AB080370.1 | - | 2 | 234 | ||
AK021335.1 | - | 3 | 278 | |||
AK077639.1 | - | 3 | 280 | |||
BC016594.1 | - | 3 | 264 | 2631627 | ||
CB236103.1 | - | 2 | 503 |