Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Dusp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023173.2 | NP_075662.2 | 6 | 1018 | ||
GenBank mRNA | AF268196.1 | - | 6 | 943 | ||
AF280810.1 | - | 6 | 885 | |||
AF280811.1 | - | 5 | 766 | |||
AK004488.1 | - | 6 | 900 | |||
AK081862.1 | - | 831 | ||||
AK155026.1 | - | 5 | 1898 | |||
AK186222.1 | - | 2 | 92 | |||
BC099453.1 | - | 6 | 1018 | 30938295 |