Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wwtr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168281.1 | NP_001161753.1 | 2 | 358 | ||
NM_133784.3 | NP_598545.2 | 2 | 437 | |||
GenBank mRNA | AJ299430.1 | - | 2 | 372 | ||
AK009983.1 | - | 2 | 370 | |||
AK011504.1 | - | 2 | 413 | |||
AK142299.1 | - | 2 | 359 | |||
AK142720.1 | - | 2 | 362 | |||
BC004640.1 | - | 2 | 358 | 3157246 | ||
BC014727.1 | - | 2 | 389 | 4487316 |