Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabif | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145510.1 | NP_663485.1 | 2 | 380 | ||
GenBank mRNA | AK087547.1 | - | 2 | 375 | ||
AK133528.1 | - | 2 | 401 | |||
AK155054.1 | - | 2 | 380 | |||
AK156126.1 | - | 292 | ||||
AK158129.1 | - | 393 | ||||
AK165748.1 | - | 2 | 398 | |||
BC011166.1 | - | 2 | 380 | 4206804 | ||
BC024808.1 | - | 2 | 370 | 5356768 |