Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd3b7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040684.1 | NP_001035774.1 | 5 | 535 | ||
NM_133943.2 | NP_598704.2 | 5 | 587 | |||
GenBank mRNA | AF277718.1 | - | 5 | 523 | ||
AK077476.1 | - | 4 | 638 | |||
BC006822.1 | - | 4 | 598 | |||
BC012954.1 | - | 4 | 388 | |||
BC018525.1 | - | 4 | 598 | |||
BC132605.1 | - | 5 | 521 | 40130882 | ||
BC138589.1 | - | 5 | 521 | 8861609 | ||
CB951288.1 | - | 2 | 144 |
Gene Symbol: 9430064I24Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK034940.1 | - | 1 | 702 | ||
AK161951.1 | - | 1 | 702 |