Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsd3b7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040684.1 | NP_001035774.1 | 7 | 816 | ||
NM_133943.2 | NP_598704.2 | 7 | 868 | |||
GenBank mRNA | AF277718.1 | - | 7 | 804 | ||
AK077476.1 | - | 6 | 919 | |||
AK150786.1 | - | 2 | 240 | |||
BC006822.1 | - | 5 | 1266 | |||
BC012954.1 | - | 6 | 669 | |||
BC018525.1 | - | 5 | 1266 | |||
BC132605.1 | - | 7 | 802 | 40130882 | ||
BC138589.1 | - | 7 | 802 | 8861609 | ||
CB951288.1 | - | 4 | 425 |