Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271347.1 | NP_001258276.1 | 10 | 1630 | ||
NM_001271348.1 | NP_001258277.1 | 10 | 1591 | |||
NM_001271349.1 | NP_001258278.1 | 9 | 1528 | |||
NM_134015.3 | NP_598776.1 | 11 | 1693 | |||
GenBank mRNA | AB093260.1 | - | 9 | 1335 | ||
AK041532.1 | - | 10 | 1423 | |||
AK149139.1 | - | 10 | 1413 | |||
AK152181.1 | - | 10 | 1353 | |||
AK160086.1 | - | 9 | 1329 | |||
AK168667.1 | - | 9 | 1345 | |||
AY038079.1 | - | 11 | 1357 | |||
BC008552.1 | - | 4 | 426 | |||
BC034261.1 | - | 11 | 1373 | 4237375 |