Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smg6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002764.1 | NP_001002764.1 | 2 | 1528 | ||
GenBank mRNA | AK129203.1 | - | 1 | 147 | ||
BC047279.1 | - | 2 | 1539 | |||
BC066040.1 | - | 2 | 1528 | 6410245 | ||
BC077715.1 | - | 2 | 1491 |