Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smg6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002764.1 | NP_001002764.1 | 19 | 4220 | ||
GenBank mRNA | AK032934.1 | - | 1 | 1635 | ||
AK052642.1 | - | 2 | 326 | |||
AK129203.1 | - | 18 | 2839 | |||
AK145229.1 | - | 3 | 706 | |||
BC037179.1 | - | 17 | 2510 | |||
BC047279.1 | - | 19 | 4232 | |||
BC066040.1 | - | 19 | 4220 | 6410245 | ||
BC077715.1 | - | 19 | 4183 |