Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmed4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134020.1 | NP_598781.1 | 3, 4 | 385 | ||
GenBank mRNA | AF368921.1 | - | 3, 4 | 416 | ||
AK078425.1 | - | 3, 4 | 396 | |||
AK158876.1 | - | 3, 4 | 418 | |||
AK204643.1 | - | 1, 2 | 80 | |||
BC023041.1 | - | 3, 4 | 385 | 5360236 |