Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019435.4 | NP_062308.2 | 1 | 464 | ||
GenBank mRNA | AK028121.1 | - | 1 | 185 | ||
BC027265.1 | - | 1 | 457 | 3589704 | ||
BC096047.1 | - | 1 | 160 | |||
DV654320.1 | - | 1 | 350 | |||
Y08702.1 | - | 1 | 157 |