Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Etv5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023794.2 | NP_076283.2 | 3 | 198 | ||
GenBank mRNA | AK018368.1 | - | 3 | 250 | ||
AK031452.1 | - | 3 | 245 | |||
AK049234.1 | - | 3 | 247 | |||
AK080674.1 | - | 2 | 121 | |||
AK164091.1 | - | 3 | 247 | |||
AK168881.1 | - | 3 | 246 | |||
AY004174.1 | - | 2 | 75 | |||
BC034680.1 | - | 3 | 198 | 4036564 |