Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cabin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172549.3 | NP_766137.2 | 10 | 1695 | ||
GenBank mRNA | BC057551.1 | - | 10 | 1623 | ||
BC150825.1 | - | 9 | 1259 | 9087736 |