Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhox9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023894.1 | NP_076383.1 | 2 | 398 | ||
GenBank mRNA | AF201698.1 | - | 2 | 398 | ||
BC099451.1 | - | 2 | 337 | 30716051 | ||
BC099963.1 | - | 2 | 332 | 30083999 | ||
DQ058646.1 | - | 2 | 398 |
Gene Symbol: Rhox6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008955.1 | NP_032981.1 | 2 | 350 | ||
GenBank mRNA | AF017453.1 | - | 2 | 350 | ||
AK076186.1 | - | 2 | 359 | |||
BC099477.1 | - | 2 | 346 | 30790014 | ||
DQ058643.1 | - | 2 | 350 |