Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnxl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144868.3 | NP_659117.2 | 1 | 855 | ||
GenBank mRNA | AF237953.1 | - | 1 | 96 | ||
AK171516.1 | - | 1 | 642 | |||
BC054751.1 | - | 1 | 134 | |||
BC055289.1 | - | 2 | 658 | |||
BC096538.1 | - | 1 | 855 |