Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc117 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134033.2 | NP_598794.2 | 4 | 709 | ||
GenBank mRNA | BC003203.1 | - | 2 | 132 | ||
BC018601.1 | - | 4 | 679 | |||
BC034558.1 | - | 4 | 647 | 3676182 | ||
BC059888.1 | - | 4 | 664 | 6816177 |