Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930427A07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134041.3 | NP_598802.2 | 7 | 863 | ||
GenBank mRNA | AK029651.1 | - | 7 | 865 | ||
AK146189.1 | - | 860 | ||||
AK167092.1 | - | 7 | 822 | |||
AK220305.1 | - | 951 | ||||
BC022617.1 | - | 4 | 437 | |||
BC116830.1 | - | 3 | 202 | |||
BC116834.1 | - | 3 | 202 |