Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps37c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181403.2 | NP_852068.1 | 5 | 674 | ||
GenBank mRNA | AK030717.1 | - | 4 | 545 | ||
AK150606.1 | - | 5 | 655 | |||
AK158833.1 | - | 4 | 413 | |||
AK159309.1 | - | 5 | 747 | |||
BC025865.1 | - | 5 | 674 | 5029143 |