Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pi4kb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293715.1 | NP_001280644.1 | 2 | 895 | ||
NM_175356.3 | NP_780565.2 | 2 | 895 | |||
GenBank mRNA | AK028974.1 | - | 2 | 880 | ||
AK030704.1 | - | 2 | 655 | |||
AK053614.1 | - | 2 | 890 | |||
AK157918.1 | - | 2 | 687 | |||
AK172074.1 | - | 2 | 888 | |||
BC079846.1 | - | 2 | 895 | 5707855 | ||
BC113176.1 | - | 2 | 750 | 40060773 | ||
BC138457.1 | - | 2 | 798 | 8861477 | ||
BC138458.1 | - | 2 | 798 | 8861478 |