Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bfsp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002896.2 | NP_001002896.1 | 5 | 1066 | ||
GenBank mRNA | AK042302.1 | - | 1 | 1939 | ||
AK134810.1 | - | 5 | 998 | |||
AK178846.1 | - | 1 | 50 | |||
AK195636.1 | - | 1 | 50 | |||
BC062815.1 | - | 3 | 268 | |||
BC068172.1 | - | 5 | 1066 | 30550909 | ||
S55549.1 | - | 3 | 375 |