Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ap4e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175550.3 | NP_780759.2 | 18 | 2645 | ||
GenBank mRNA | AK036944.1 | - | 18 | 2626 | ||
AK144636.1 | - | 18 | 2626 | |||
BC042530.1 | - | 5 | 836 |