Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prkab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182997.2 | NP_892042.2 | 4 | 580 | ||
GenBank mRNA | BC060228.1 | - | 4 | 580 | 6839848 |