Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175375.3 | NP_780584.2 | 20 | 3765 | ||
GenBank mRNA | AK017018.1 | - | 10 | 1104 | ||
AK029322.1 | - | 13 | 2180 | |||
AK042754.1 | - | 7 | 740 | |||
AK131166.1 | - | 20 | 3628 |