Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnaaf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027269.3 | NP_081545.3 | 1 | 1636 | ||
GenBank mRNA | AB455811.1 | - | 1 | 1492 | ||
AK012767.1 | - | 1 | 314 | |||
AK075648.1 | - | 288 | ||||
AK154254.1 | - | 1 | 341 | |||
AK172494.1 | - | 1 | 312 | |||
BC055807.1 | - | 1 | 1636 | 6414041 | ||
BC058344.1 | - | 221 | ||||
BC062898.1 | - | 1 | 1636 | 6414041 | ||
EU599049.1 | - | 311 |