Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6ka3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_148945.2 | NP_683747.1 | 15 | 1590 | ||
GenBank mRNA | AK079102.1 | - | 1683 | |||
AK154778.1 | - | 15 | 1502 | |||
AY083469.1 | - | 15 | 1252 | |||
BC150156.1 | - | 15 | 1255 | |||
BC150478.1 | - | 15 | 1255 |