Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077190.2 | NP_001070658.1 | 2 | 336 | ||
NM_001077192.2 | NP_001070660.1 | 2 | 336 | |||
NM_001077193.2 | NP_001070661.1 | 2 | 336 | |||
NM_007380.3 | NP_031406.2 | 2 | 336 | |||
NM_145994.2 | NP_666106.1 | 2 | 336 | |||
GenBank mRNA | AF420251.1 | - | 2 | 272 | ||
AK034476.1 | - | 2 | 218 | |||
AK151026.1 | - | 2 | 244 | |||
AK151363.1 | - | 2 | 315 | |||
AK152061.1 | - | 2 | 200 | |||
AK152184.1 | - | 2 | 200 | |||
AK153130.1 | - | 2 | 244 | |||
AK164040.1 | - | 2 | 252 | |||
AK165924.1 | - | 2 | 309 | |||
AK167328.1 | - | 2 | 295 | |||
AK167568.1 | - | 2 | 291 | |||
AK167744.1 | - | 2 | 283 | |||
AY033645.1 | - | 2 | 180 | |||
BC004657.1 | - | 2 | 287 | 3498068 | ||
BC079642.1 | - | 2 | 242 | 6816252 | ||
BY278454.1 | - | 2 | 336 |