Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029836.2 | NP_001025007.1 | 6 | 850 | ||
NM_001287101.1 | NP_001274030.1 | 8 | 994 | |||
NM_001287102.1 | NP_001274031.1 | 7 | 943 | |||
NM_001287103.1 | NP_001274032.1 | 6 | 568 | |||
NM_033525.3 | NP_277060.2 | 7 | 901 | |||
GenBank mRNA | AB059656.1 | - | 6 | 850 | ||
AF397007.1 | - | 7 | 695 | |||
AF397008.1 | - | 8 | 746 | |||
AK050484.1 | - | 6 | 754 | |||
AK158764.1 | - | 6 | 570 | |||
AY035898.1 | - | 6 | 725 | |||
AY035899.1 | - | 7 | 776 | |||
BC046642.1 | - | 1 | 37 | |||
BC068308.1 | - | 6 | 719 | 30472074 |