Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pawr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054056.2 | NP_473397.1 | 6 | 1065 | ||
GenBank mRNA | AF377871.1 | - | 5 | 468 | ||
AK136412.1 | - | 1 | 30 | |||
DQ363525.1 | - | 6 | 1065 | |||
DQ449073.1 | - | 4 | 744 | |||
DQ449074.1 | - | 5 | 876 |