Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dscaml1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081270.1 | NP_001074739.1 | 13 | 2769 | ||
GenBank mRNA | AB093285.1 | - | 8 | 1627 | ||
AF487347.1 | - | 13 | 2661 | |||
AK147531.1 | - | 13 | 2713 | |||
BC096527.1 | - | 5 | 603 |
Set Membership: |
![]() |