Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dscaml1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081270.1 | NP_001074739.1 | 21 | 4009 | ||
GenBank mRNA | AB093285.1 | - | 16 | 2867 | ||
AF351196.1 | - | 3 | 303 | |||
AF487348.1 | - | 7 | 957 | |||
AF487349.1 | - | 5 | 794 | |||
AK147531.1 | - | 21 | 3954 | |||
BC096527.1 | - | 13 | 1843 |
Set Membership: |
![]() |