Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dscaml1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081270.1 | NP_001074739.1 | 3 | 641 | ||
GenBank mRNA | AF487345.1 | - | 3 | 545 | ||
AF487347.1 | - | 3 | 533 | |||
AK014832.1 | - | 787 | ||||
AK015320.1 | - | 802 | ||||
AK147531.1 | - | 3 | 585 | |||
AK161365.1 | - | 670 |
Set Membership: |
![]() |