Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crisp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009639.2 | NP_033769.1 | 707 | |||
GenBank mRNA | BC022573.1 | - | 8 | 728 | 4925349 | |
BC114345.2 | - | 709 | ||||
BC132536.1 | - | 706 | 40130813 | |||
BC132538.1 | - | 706 | 40130815 | |||
L05560.1 | - | 707 | ||||
M92850.1 | - | 8 | 715 |