- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: Apoh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013475.4 | NP_038503.4 | 5 | 494 | ||
GenBank mRNA | AA273732.1 | - | 2 | 109 | ||
AK010926.1 | - | 5 | 482 | |||
BC011037.1 | - | 5 | 510 | |||
BC019811.1 | - | 5 | 482 | 5125770 | ||
BC053338.1 | - | 5 | 470 | 5125963 | ||
D10056.1 | - | 5 | 468 | |||
S70439.1 | - | 5 | 478 | |||
Y11356.1 | - | 5 | 498 |