Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Art1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009710.4 | NP_033840.2 | 3 | 817 | ||
GenBank mRNA | AK003107.1 | - | 3 | 818 | ||
AK009902.1 | - | 818 | ||||
BC063760.1 | - | 3 | 798 | 30137479 | ||
BC132492.1 | - | 3 | 805 | 40130769 | ||
BC132496.1 | - | 3 | 805 | 40130773 | ||
X95842.1 | - | 2 | 779 |