Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007505.2 | NP_031531.1 | 4 | 625 | ||
GenBank mRNA | AK043976.1 | - | 4 | 393 | ||
AK076572.1 | - | 4 | 393 | |||
AK098155.1 | - | 4 | 403 | |||
AK146797.1 | - | 4 | 396 | |||
AK150426.1 | - | 4 | 396 | |||
AK150843.1 | - | 4 | 393 | |||
AK151004.1 | - | 4 | 393 | |||
AK151128.1 | - | 4 | 402 | |||
AK151224.1 | - | 4 | 400 | |||
AK151920.1 | - | 4 | 397 | |||
AK152054.1 | - | 4 | 404 | |||
AK152890.1 | - | 4 | 396 | |||
AK153167.1 | - | 4 | 397 | |||
AK153282.1 | - | 4 | 393 | |||
AK157529.1 | - | 2 | 174 | |||
AK159491.1 | - | 4 | 393 | |||
AK159540.1 | - | 4 | 393 | |||
AK159758.1 | - | 4 | 396 | |||
AK160043.1 | - | 4 | 403 | |||
AK164110.1 | - | 4 | 396 | |||
AK164193.1 | - | 2 | 99 | |||
AK166709.1 | - | 4 | 396 | |||
AK166812.1 | - | 4 | 409 | |||
AK167159.1 | - | 4 | 393 | |||
AK167863.1 | - | 4 | 373 | |||
AK168198.1 | - | 4 | 403 | |||
AK168369.1 | - | 4 | 393 | |||
AK168617.1 | - | 4 | 393 | |||
AK168879.1 | - | 4 | 393 | |||
AK168890.1 | - | 4 | 393 | |||
AK168932.1 | - | 4 | 393 | |||
AK169080.1 | - | 4 | 396 | |||
AK169084.1 | - | 4 | 396 | |||
AK169105.1 | - | 4 | 393 | |||
AK169142.1 | - | 4 | 438 | |||
AK169300.1 | - | 4 | 393 | |||
AK169308.1 | - | 4 | 398 | |||
AK169414.1 | - | 4 | 396 | |||
AK192613.1 | - | 2 | 152 | |||
AK196974.1 | - | 2 | 152 | |||
AK209959.1 | - | 2 | 152 | |||
AK218124.1 | - | 4 | 322 | |||
BC014854.1 | - | 4 | 380 | 4500893 | ||
BC020417.1 | - | 4 | 380 | |||
BC055892.1 | - | 4 | 830 | |||
L01062.1 | - | 4 | 380 |